Learn More
System Biosciences LLC EXOGLOW-NTA FLO LBL KIT ZETA
SDP
Catalog No. NC1528815
Encompass_Preferred
See only extracellular vesicles and exosomes for more accurate particle size analysis and quantitation using Particle Metrix's ZetaView. Building upon ExoGlow-NTA, a proprietary dye that enables fluorescent NTA analysis of only the extracellular vesicles present in a heterogenous sample, SBI has developed a new version compatible with Particle Metrix's ZetaView instrument. The new ExoGlow-NTA Fluorescent Labeling Kit (for Particle Metrix ZetaView) provides accurate EV NTA data that excludes protein aggregates, membrane fractions, and other background particles to provide EV-specific particle size and concentration.
Catalog No. NC1528815 Supplier System Biosciences LLC Supplier No. EXONTA110A1
See only extracellular vesicles and exosomes for more accurate particle size analysis and quantitation using Particle Metrix's ZetaView. Building upon ExoGlow-NTA, a proprietary dye that enables fluorescent NTA analysis of only the extracellular vesicles present in a heterogenous sample, SBI has developed a new version compatible with Particle Metrix's ZetaView instrument. The new ExoGlow-NTA Fluorescent Labeling Kit (for Particle Metrix ZetaView) provides accurate EV NTA data that excludes protein aggregates, membrane fractions, and other background particles to provide EV-specific particle size and concentration.