Learn More
System Biosciences LLC EXOGLOW PROTEIN EV LABL K/GRNExoGlowTM-Protein EV Labeling Kit (Green) = 20 Reactions
SDP
Catalog No. NC1528816
Encompass_Preferred
Label EV proteins green (511 nm excitation/525 nm emission) with a reagent specifically optimized for Exosome labeling. SBI’s new generation of extracellular vesicle (EV) labeling reagents take your EV visualization to new levels of clarity, with low background and high selectivity. Unlike general-purpose labeling reagents that are not optimized for EVs and suffer from high levels of background signal, ExoGlow™-Protein Green improves your ability to track and localize EVs by specifically and covalently labeling internal EV proteins with a proprietary green fluorescent dye that delivers very low levels of background signal. The result is unmatched EV imaging for more accurate studies (see below for our full range of next generation ExoGlow reagents).
Catalog No. NC1528816 Supplier System Biosciences LLC Supplier No. EXOGP300A1
Label EV proteins green (511 nm excitation/525 nm emission) with a reagent specifically optimized for Exosome labeling. SBI’s new generation of extracellular vesicle (EV) labeling reagents take your EV visualization to new levels of clarity, with low background and high selectivity. Unlike general-purpose labeling reagents that are not optimized for EVs and suffer from high levels of background signal, ExoGlow™-Protein Green improves your ability to track and localize EVs by specifically and covalently labeling internal EV proteins with a proprietary green fluorescent dye that delivers very low levels of background signal. The result is unmatched EV imaging for more accurate studies (see below for our full range of next generation ExoGlow reagents).