Learn More
New England Biolabs, Inc. Exonuclease I (E.coli) – 15000 units
SDP
Catalog No. 50811715
Encompass
Exonuclease I is a DNA-specific endonuclease that catalyzes the removal of nucleotides from linear single-stranded DNA in the 3' to 5' direction. Note: See also Exo-CIP Rapid PCR Cleanup Kit.
Catalog No. 50-811-715 Supplier New England Biolabs, Inc. Supplier No. M0293L
Exonuclease I is a DNA-specific endonuclease that catalyzes the removal of nucleotides from linear single-stranded DNA in the 3' to 5' direction. Note: See also Exo-CIP Rapid PCR Cleanup Kit.