Learn More
New England Biolabs, Inc. Exonuclease I (E.coli) – 3000 units
SDP
Catalog No. 50811716
Encompass
Exonuclease I is a DNA-specific endonuclease that catalyzes the removal of nucleotides from linear single-stranded DNA in the 3' to 5' direction. Note: See also Exo-CIP Rapid PCR Cleanup Kit.
Catalog No. 50-811-716 Supplier New England Biolabs, Inc. Supplier No. M0293S
Exonuclease I is a DNA-specific endonuclease that catalyzes the removal of nucleotides from linear single-stranded DNA in the 3' to 5' direction. Note: See also Exo-CIP Rapid PCR Cleanup Kit.