Learn More
Agilent Technologies EXO-KLENOW
Supplier:  Agilent Technologies 600069
Encompass
Exo- Klenow is a exonuclease-deficient version of DNA polymerase I exhibiting 5' to 3' DNA polymerase activity. It was isolated from an overproducing recombinant E. coli strain and is used for sequencing and labeling applications.
Catalog No. 50-125-900
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.