Learn More
Mayflower Bioscience Exo-spin 20 mini-HD columns
SDP
Catalog No. 501979300
Encompass
Size exclusion chromatography (SEC) columns are a reliable method for exosome isolation and can be used as a stand-alone product or as part of a combined strategy for isolating small EVs. The EX05 Exo-spin™ kit contains packed and equilibrated ready-to-use SEC columns.
Catalog No. 50-197-9300 Supplier Mayflower Bioscience Supplier No. EX0520
Size exclusion chromatography (SEC) columns are a reliable method for exosome isolation and can be used as a stand-alone product or as part of a combined strategy for isolating small EVs. The EX05 Exo-spin™ kit contains packed and equilibrated ready-to-use SEC columns.