Learn More
CELL GUIDANCE SYSTEMS LLC EXO-SPIN 96
SDP
Catalog No. NC2145507
Encompass
Exo-spin™ 96 provides proven size exclusion chromatography (SEC) mini-column (EX01) technology in a 96-well format, allowing up to 96 individual samples to be processed in less than 30 minutes. Sample volumes up to 0.5 ml can be purified. The kit provides flexibility with 8-column strips detachable from the plate, so you can process 8 samples in 8-column strips if neededThe Exo-spin™ 96 well product offers:High-throughput exosome isolation at the benchtop in the standard 96 well format in under 30 minutesPurify up to 0.5 ml of each sampleUp to 99% of free proteins removedOutstanding reproducibility +/- 5% Standard deviation, for consistent exosome purification, time after timeFlexibility to just work with single strips of 8 columns at a timeThe convenience of purified exosomes eluted directly into a 96 well plate ready for high throughput downstream applications
This item is not returnable. View return policy
Catalog No. NC2145507 Supplier CELL GUIDANCE SYSTEMS LLC Supplier No. EX0796
Exo-spin™ 96 provides proven size exclusion chromatography (SEC) mini-column (EX01) technology in a 96-well format, allowing up to 96 individual samples to be processed in less than 30 minutes. Sample volumes up to 0.5 ml can be purified. The kit provides flexibility with 8-column strips detachable from the plate, so you can process 8 samples in 8-column strips if neededThe Exo-spin™ 96 well product offers:High-throughput exosome isolation at the benchtop in the standard 96 well format in under 30 minutesPurify up to 0.5 ml of each sampleUp to 99% of free proteins removedOutstanding reproducibility +/- 5% Standard deviation, for consistent exosome purification, time after timeFlexibility to just work with single strips of 8 columns at a timeThe convenience of purified exosomes eluted directly into a 96 well plate ready for high throughput downstream applications