Learn More
Mayflower Bioscience Exo-spin blood, 8 columns, 60 ml buffer
SDP
Catalog No. 501979291
Encompass
The EX02 Exo-spin™ kit has been developed to process small volumes of blood samples (both plasma and sera). The kit includes Exo-spin™ buffer as well as SEC columns. The Exo-spin™ buffer is a polymer-based precipitation buffer and will be used for the first step. For the second step, pre-packed and equilibrated ready-to-use SEC columns are included.
Catalog No. 50-197-9291 Supplier Mayflower Bioscience Supplier No. EX028
The EX02 Exo-spin™ kit has been developed to process small volumes of blood samples (both plasma and sera). The kit includes Exo-spin™ buffer as well as SEC columns. The Exo-spin™ buffer is a polymer-based precipitation buffer and will be used for the first step. For the second step, pre-packed and equilibrated ready-to-use SEC columns are included.