Learn More
New England Biolabs, Inc. Exonuclease T – 250 units
SDP
Catalog No. 50811693
Encompass
Exonuclease T (Exo T), also known as RNase T, is a single-stranded RNA or DNA specific nuclease that requires a free 3' terminus and removes nucleotides in the 3' to 5' direction. Exonuclease T can be used to generate blunt ends from RNA or DNA molecules that have 3 extensions.
Catalog No. 50-811-693 Supplier New England Biolabs, Inc. Supplier No. M0265S
Exonuclease T (Exo T), also known as RNase T, is a single-stranded RNA or DNA specific nuclease that requires a free 3' terminus and removes nucleotides in the 3' to 5' direction. Exonuclease T can be used to generate blunt ends from RNA or DNA molecules that have 3 extensions.