Learn More
System Biosciences LLC EXO URINE EV ISOLATION KITExo-Urine™ EV Isolation Kit = 10 Reactions
SDP
Catalog No. NC2229335
Encompass_Preferred
High yields and high purity EVs from urine—SBI’s Exo-Urine™ EV Isolation Kit overcomes the challenges of conventional Exosome isolation. Overcome the yield and purity challenges of isolating EVs from urine with SBI’s Exo-Urine EV Isolation Kit. Designed to help EVs escape from the mesh of polymerized Tamm-Horstall protein (THP/uromodulin), a glycoprotein present in high amounts in urine, the Exo-Urine EV Isolation Kit combines size-exclusion chromatography (SEC) and proprietary solubilization reagents to deliver high yields of highly pure EVs. Starting with as little as 1.5 mL of freshly voided (<4 hrs from initial collection) or frozen urine preps, the Exo-Urine Kit delivers EV preps that are low in THP/uromodulin contamination, enabling faster, more efficient studies of EV-derived biomarkers from urine..
This item is not returnable. View return policy
Catalog No. NC2229335 Supplier System Biosciences LLC Supplier No. EXOU100A1
High yields and high purity EVs from urine—SBI’s Exo-Urine™ EV Isolation Kit overcomes the challenges of conventional Exosome isolation. Overcome the yield and purity challenges of isolating EVs from urine with SBI’s Exo-Urine EV Isolation Kit. Designed to help EVs escape from the mesh of polymerized Tamm-Horstall protein (THP/uromodulin), a glycoprotein present in high amounts in urine, the Exo-Urine EV Isolation Kit combines size-exclusion chromatography (SEC) and proprietary solubilization reagents to deliver high yields of highly pure EVs. Starting with as little as 1.5 mL of freshly voided (<4 hrs from initial collection) or frozen urine preps, the Exo-Urine Kit delivers EV preps that are low in THP/uromodulin contamination, enabling faster, more efficient studies of EV-derived biomarkers from urine..