Learn More
Aviva Systems Biology Corp EXO1 Antibody - C-terminal region (ARP63489_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63489_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EXO1. It was validated on Western Blot. . Alternate names: HEX1, hExoIApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-986