Learn More
System Biosciences LLC EXOAB ANTIBODY KIT CD9 CD63ExoAb Antibody Kit (CD9, CD63, CD81, Hsp70 antibodies, rabbit anti-human) with goat anti-rabbit HRP secondary antibody = 25ul Each
SDP
Catalog No. NC0256458
Encompass_Preferred
Be confident in your exosome detection with this kit that includes antibodies to the most commonly-used exosome markers CD9, CD63, CD81, and HSP70. When you’re ready to confirm that you’ve isolated exosomes, stick with the exosome experts at SBI. Our ExoAb Antibody Kit includes antibodies to four of the most commonly used exosome markers—CD9, CD63, CD81, and HPS70. These are all polyclonal rabbit antibodies that recognize the human forms of the protein, and are a great choice for general exosome detection.
Catalog No. NC0256458 Supplier System Biosciences LLC Supplier No. EXOABKIT1
Be confident in your exosome detection with this kit that includes antibodies to the most commonly-used exosome markers CD9, CD63, CD81, and HSP70. When you’re ready to confirm that you’ve isolated exosomes, stick with the exosome experts at SBI. Our ExoAb Antibody Kit includes antibodies to four of the most commonly used exosome markers—CD9, CD63, CD81, and HPS70. These are all polyclonal rabbit antibodies that recognize the human forms of the protein, and are a great choice for general exosome detection.