Learn More
System Biosciences LLC EXOBACTERIA OMV ISOLATION KIT - 20 Reactions
SDP
Catalog No. NC1657922
Encompass_Preferred
Get clean, high-yield preps of bacterial outer membrane vesicles (OMVs) from an easy-to-use workflow. Expanding our exosome expertise to bacteria, SBI now offers the ExoBacteria™ OMV Isolation Kit. The first and only kit designed to streamline isolation of bacterial outer membrane vesicles (OMVs), the ExoBacteria OMV Isolation Kit uses an innovative, precipitation-free gravity column system to harvest OMVs from bacterial culture medium.
Catalog No. NC1657922 Supplier System Biosciences LLC Supplier No. EXOBAC100A1
Get clean, high-yield preps of bacterial outer membrane vesicles (OMVs) from an easy-to-use workflow. Expanding our exosome expertise to bacteria, SBI now offers the ExoBacteria™ OMV Isolation Kit. The first and only kit designed to streamline isolation of bacterial outer membrane vesicles (OMVs), the ExoBacteria OMV Isolation Kit uses an innovative, precipitation-free gravity column system to harvest OMVs from bacterial culture medium.