Learn More
Aviva Systems Biology Corp EXOC1 Antibody - N-terminal region (ARP57769_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57769_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EXOC1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human EXOC1. Alternate names: BM-102, FLJ10893, SEC3, SEC3L1, SEC3PApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-372