Learn More
ABclonal Technology EXOC3 Rabbit mAb
SDP
Catalog No. 5000007921
Encompass_Preferred
The protein encoded by this gene is a component of the exocyst complex a multiple protein complex essential for targeting exocytic vesicles to specific docking sites on the plasma membrane Though best characterized in yeast the component proteins and functions of exocyst complex have been demonstrated to be highly conserved in higher eukaryotes At least eight components of the exocyst complex including this protein are found to interact with the actin cytoskeletal remodeling and vesicle transport machinery The complex is also essential for the biogenesis of epithelial cell surface polarity
Catalog No. 50-000-07921 Supplier ABclonal Technology Supplier No. A20925100UL
The protein encoded by this gene is a component of the exocyst complex a multiple protein complex essential for targeting exocytic vesicles to specific docking sites on the plasma membrane Though best characterized in yeast the component proteins and functions of exocyst complex have been demonstrated to be highly conserved in higher eukaryotes At least eight components of the exocyst complex including this protein are found to interact with the actin cytoskeletal remodeling and vesicle transport machinery The complex is also essential for the biogenesis of epithelial cell surface polarity