Learn More
Aviva Systems Biology Corp EXOC4 Antibody - N-terminal region (ARP56254_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56254_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EXOC4. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human EXOC4. Alternate names: MGC27170, REC8, SEC8, SEC8L1, Sec8pApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-286-131