Learn More
Aviva Systems Biology Corp EXOC6 Antibody - N-terminal region (ARP57319_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57319_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EXOC6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human EXOC6. Alternate names: DKFZp761I2124, EXOC6A, FLJ1125, FLJ11251, MGC33397, SEC15L, SEC15L1, SEC15L3, Sec15p, SEC15ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53952