Learn More
System Biosciences LLC EXOCET EXOSOME QUANT KIT = 9 Reactions
SDP
Catalog No. NC0894393
Encompass_Preferred
Find out how many extracellular vesicles you have in just 20 minutes with this antibody-free, colorimetric assay. With the EXOCET Exosome Quantitation Kit, you can find out just how many exosomes you’ve isolated in as little as 20 minutes. This antibody-free, colorimetric assay (OD405) measures the activity of Acetyl-CoA Acetylcholinesterase (AChE), an enzyme shown to be highly enriched in exosomes1, 2 (using mass spectrometry and Western blotting, SBI has verified the presence of AChE in exosomes from serum, stem cells, cancer cell lines, and MSCs
Catalog No. NC0894393 Supplier System Biosciences LLC Supplier No. EXOCET96A1
Find out how many extracellular vesicles you have in just 20 minutes with this antibody-free, colorimetric assay. With the EXOCET Exosome Quantitation Kit, you can find out just how many exosomes you’ve isolated in as little as 20 minutes. This antibody-free, colorimetric assay (OD405) measures the activity of Acetyl-CoA Acetylcholinesterase (AChE), an enzyme shown to be highly enriched in exosomes1, 2 (using mass spectrometry and Western blotting, SBI has verified the presence of AChE in exosomes from serum, stem cells, cancer cell lines, and MSCs