Learn More
System Biosciences LLC EXOCET-STANDARDS = 1 set
SDP
Catalog No. NC2171489
Encompass_Preferred
Standards for our fast and easy EXOCET assay - Exosome Quantitation. While our fast and easy EXOCET Kit comes with standards, we also offer our EXOCET exosome standards as a stand-alone product for when you need more. The standards are calibrated using NanoSight analysis, enabling estimation of the number of exosomes in your samples.
This item is not returnable. View return policy
Catalog No. NC2171489 Supplier System Biosciences LLC Supplier No. EXOCETSD1
Standards for our fast and easy EXOCET assay - Exosome Quantitation. While our fast and easy EXOCET Kit comes with standards, we also offer our EXOCET exosome standards as a stand-alone product for when you need more. The standards are calibrated using NanoSight analysis, enabling estimation of the number of exosomes in your samples.