Learn More
System Biosciences LLC EXOCHECK EXOSOME ARRAYS - 4 ARRAYSExo-Check Exosome Antibody Array = 4 Arrays
SDP
Catalog No. NC2749842
Encompass_Preferred
Exo-Check™ Exosome Antibody Array - for the most efficient detection of exosomes. For the most efficient detection of exosomes, turn to our semi-quantitative Exo-Check Exosome Antibody Arrays. Each array has 12 pre-printed spots and features 8 antibodies for known exosome markers (CD63, CD81, ALIX, FLOT1, ICAM1, EpCam, ANXA5 and TSG101), a GM130 cis-Golgi marker to monitor any cellular contamination in your exosome isolations, a labeled positive control for HRP detection, and a blank spot as a background control. The kits come complete with a secondary detection mixture conjugated to HRP.
Catalog No. NC2749842 Supplier System Biosciences LLC Supplier No. EXORAY200B4
Exo-Check™ Exosome Antibody Array - for the most efficient detection of exosomes. For the most efficient detection of exosomes, turn to our semi-quantitative Exo-Check Exosome Antibody Arrays. Each array has 12 pre-printed spots and features 8 antibodies for known exosome markers (CD63, CD81, ALIX, FLOT1, ICAM1, EpCam, ANXA5 and TSG101), a GM130 cis-Golgi marker to monitor any cellular contamination in your exosome isolations, a labeled positive control for HRP detection, and a blank spot as a background control. The kits come complete with a secondary detection mixture conjugated to HRP.