Learn More
Aviva Systems Biology Corp EXOD1 Antibody - N-terminal region (ARP52508_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52508_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EXOD1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human EXOD1. Alternate names: KIAA1504, MGC16943, EXOD1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-285-722