Learn More
System Biosciences LLC ExoELISA-ULTRA Complete Kit (CD63 detection), 96 reactions
SDP
Supplier:  System Biosciences LLC EXELULTRACD631
Encompass
With a 4-hour total assay time, this sensitive ELISA-based assay speeds quantitation of exosomes from most biofluids.
Catalog No. NC1242688