Learn More
Medchemexpress LLC Exoenzyme C3, clostridium botulinum | 58319-92-9 | 25 UG
SDP
Catalog No. 5000425858
Encompass_Preferred
Exoenzyme C3, Clostridium botulinum, is a mono-ADP-ribosylating enzyme. It specifically modifies RhoA, B, and C by transferring ADP-ribose to them, thereby inactivating these GTPases. It can induce neuronal axonal and dendritic growth, inhibit macrophage migration, and regulate cytoskeletal dynamics.
  • Mono-ADP-ribosylating enzyme
  • Specifically modifies RhoA, B, and C
  • Induces neuronal axonal and dendritic growth
  • Inhibits macrophage migration
  • Regulates cytoskeletal dynamics
  • Used in research for spinal cord injury
  • Used in research for diabetic painful neuropathy
Catalog No. 50-004-25858 Supplier Medchemexpress LLC Supplier No. 25UGHYP2325
Exoenzyme C3, Clostridium botulinum, is a mono-ADP-ribosylating enzyme. It specifically modifies RhoA, B, and C by transferring ADP-ribose to them, thereby inactivating these GTPases. It can induce neuronal axonal and dendritic growth, inhibit macrophage migration, and regulate cytoskeletal dynamics.
  • Mono-ADP-ribosylating enzyme
  • Specifically modifies RhoA, B, and C
  • Induces neuronal axonal and dendritic growth
  • Inhibits macrophage migration
  • Regulates cytoskeletal dynamics
  • Used in research for spinal cord injury
  • Used in research for diabetic painful neuropathy