Learn More
Abcam E x oFACS Kit, 20 Tests
Catalog No. 502679994
Encompass_Preferred
ExoFACS Kit ab287876 (previously known as ExoFACS kit for Cell Culture Media Exosomes K1236) allows exosome isolation from biofluids or cell culture media and FACS analysis of exosome markers.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-9994 Supplier Abcam Supplier No. AB28787620TEST
ExoFACS Kit ab287876 (previously known as ExoFACS kit for Cell Culture Media Exosomes K1236) allows exosome isolation from biofluids or cell culture media and FACS analysis of exosome markers.
The product is subject to the following: Abcam Restricted Use Statement