Learn More
MSC EXOFIT XP ARC FLASH HRNS
Catalog No. 5000802234
Encompass
The ExoFit XP Safety Harness is designed for arc-flash applications, incorporating premium comfort padding that is both removable and washable, continuing the legacy of the original ExoFit Comfort Harnesses.
  • Automatic stand-up back D-ring for efficient dorsal connections
  • Removable shoulder, back, and leg comfort padding
  • Protected labels equipped with an integrated RFID tag for inventory, inspection, and record management
Catalog No. 50-008-02234 Supplier MSC Supplier No. 64285232
The ExoFit XP Safety Harness is designed for arc-flash applications, incorporating premium comfort padding that is both removable and washable, continuing the legacy of the original ExoFit Comfort Harnesses.
  • Automatic stand-up back D-ring for efficient dorsal connections
  • Removable shoulder, back, and leg comfort padding
  • Protected labels equipped with an integrated RFID tag for inventory, inspection, and record management