Learn More
System Biosciences LLC EXOFLOW-ONE EV LABELING KIT GFRExoFlow-ONE EV Labeling Kit for Flow Cytometry, Garnet Far Red = 25 Reactions
SDP
Catalog No. NC1630932
Encompass_Preferred
Finally, you can take full advantage of the power of flow-based methods for analyzing extracellular vesicles (EVs) with Directly detect extracellular vesicles down to near single-vesicle resolution for more powerful insights from flow cytometry applications. SBI’s exclusive ExoFlow-ONE Gemstone dyes. By specifically labeling internal EV components with one of these proprietary, high quantum efficiency dyes, you can achieve near single-vesicle resolution* for more powerful flow cytometry and FACS studies, and uncover greater insights into EV biology.
Catalog No. NC1630932 Supplier System Biosciences LLC Supplier No. EXOF200A1
Finally, you can take full advantage of the power of flow-based methods for analyzing extracellular vesicles (EVs) with Directly detect extracellular vesicles down to near single-vesicle resolution for more powerful insights from flow cytometry applications. SBI’s exclusive ExoFlow-ONE Gemstone dyes. By specifically labeling internal EV components with one of these proprietary, high quantum efficiency dyes, you can achieve near single-vesicle resolution* for more powerful flow cytometry and FACS studies, and uncover greater insights into EV biology.