Learn More
System Biosciences LLC EXOGLOW MEMBRANE EV LABELING Kit = 25 reactions
SDP
Catalog No. NC1386229
Encompass_Preferred
Label EV membranes (465 nm excitation/635 nm emission) with a reagent specifically optimized for Exosome labeling. SBI’s new generation of extracellular vesicle (EV) labeling reagents take your EV visualization to new levels of clarity, with low background and high selectivity. Unlike general-purpose labeling reagents that are not optimized for EVs and suffer from high levels of background signal, ExoGlow™-Membrane improves your ability to track and localize EVs by specifically labeling EV membranes with a proprietary fluorescent dye that delivers very low levels of background signal. The result is unmatched EV imaging for more accurate studies (see below for our full range of next generation ExoGlow reagents).
Catalog No. NC1386229 Supplier System Biosciences LLC Supplier No. EXOGM600A1
Label EV membranes (465 nm excitation/635 nm emission) with a reagent specifically optimized for Exosome labeling. SBI’s new generation of extracellular vesicle (EV) labeling reagents take your EV visualization to new levels of clarity, with low background and high selectivity. Unlike general-purpose labeling reagents that are not optimized for EVs and suffer from high levels of background signal, ExoGlow™-Membrane improves your ability to track and localize EVs by specifically labeling EV membranes with a proprietary fluorescent dye that delivers very low levels of background signal. The result is unmatched EV imaging for more accurate studies (see below for our full range of next generation ExoGlow reagents).