Learn More
System Biosciences LLC EXOGLOW-RNA EV LABELING KIT = 10 Reactions
SDP
Catalog No. NC2337886
Encompass_Preferred
Label EV mRNA (485 nm excitation/537 nm emission) with a reagent specifically optimized for Exosome labeling. SBI’s new generation of extracellular vesicle (EV) labeling reagents take your EV visualization to new levels of clarity, with low background and high selectivity. Unlike general-purpose labeling reagents that are not optimized for EVs and suffer from high levels of background signal, ExoGlow™-RNA improves your ability to track and localize EVs by specifically labeling EV mRNAs with a proprietary fluorescent dye that delivers very low levels of background signal. The result is unmatched EV imaging for more accurate EV studies (see below for our full range of next generation ExoGlow reagents).
Catalog No. NC2337886 Supplier System Biosciences LLC Supplier No. EXOGR800A1
Label EV mRNA (485 nm excitation/537 nm emission) with a reagent specifically optimized for Exosome labeling. SBI’s new generation of extracellular vesicle (EV) labeling reagents take your EV visualization to new levels of clarity, with low background and high selectivity. Unlike general-purpose labeling reagents that are not optimized for EVs and suffer from high levels of background signal, ExoGlow™-RNA improves your ability to track and localize EVs by specifically labeling EV mRNAs with a proprietary fluorescent dye that delivers very low levels of background signal. The result is unmatched EV imaging for more accurate EV studies (see below for our full range of next generation ExoGlow reagents).