Learn More
System Biosciences LLC EXOGLOW-NTA FLUORESCENTExoGlow™-NTA Fluorescent Labeling Kit (for Particle Metrix ZetaView) = 10 Reeactions
SDP
Catalog No. NC1520332
Encompass_Preferred
See only extracellular vesicles and exosomes with Nanoparticle Tracking Analysis for more accurate particle size analysis and quantitation with Malvern's NanoSight. Building upon ExoGlow-NTA, a proprietary dye that enables fluorescent NTA analysis of only the extracellular vesicles present in a heterogenous sample, SBI has developed a new version compatible with Particle Metrix's ZetaView instrument. The new ExoGlow-NTA Fluorescent Labeling Kit (for Particle Metrix ZetaView) provides accurate EV NTA data that excludes protein aggregates, membrane fractions, and other background particles to provide EV-specific particle size and concentration.
Catalog No. NC1520332 Supplier System Biosciences LLC Supplier No. EXONTA200A1
See only extracellular vesicles and exosomes with Nanoparticle Tracking Analysis for more accurate particle size analysis and quantitation with Malvern's NanoSight. Building upon ExoGlow-NTA, a proprietary dye that enables fluorescent NTA analysis of only the extracellular vesicles present in a heterogenous sample, SBI has developed a new version compatible with Particle Metrix's ZetaView instrument. The new ExoGlow-NTA Fluorescent Labeling Kit (for Particle Metrix ZetaView) provides accurate EV NTA data that excludes protein aggregates, membrane fractions, and other background particles to provide EV-specific particle size and concentration.