Learn More
Boca Scientific Inc Exontrap Cloning Vector pET01, Lyophilized DNA, 5μg
Catalog No. NC1499834
Encompass
The Exontrap vector pET01 has an intrinsic splicing function that enables selective cloning of exon sequences from large genomic eukaryotic DNA fragments. This new route for the identification of eukaryotic genes does not involve an initial isolation of cellular mRNA, allowing genes that are not transcribed during certain life cycle stages to be identified. This greatly facilitates exon/intron mapping. The Exontrap function is based on a shuttle vector containing prokaryotic and eukaryotic genetic elements for replication in both bacteria and cell cultures. The vector contains a 5´ and 3´ exon separated by a 600 bp intron sequence that has a polylinker for cloning.
Catalog No. NC1499834 Supplier Boca Scientific Inc Supplier No. PET01
The Exontrap vector pET01 has an intrinsic splicing function that enables selective cloning of exon sequences from large genomic eukaryotic DNA fragments. This new route for the identification of eukaryotic genes does not involve an initial isolation of cellular mRNA, allowing genes that are not transcribed during certain life cycle stages to be identified. This greatly facilitates exon/intron mapping. The Exontrap function is based on a shuttle vector containing prokaryotic and eukaryotic genetic elements for replication in both bacteria and cell cultures. The vector contains a 5´ and 3´ exon separated by a 600 bp intron sequence that has a polylinker for cloning.