Learn More
Sigma Aldrich Fine Chemicals Biosciences Exonuclease I 10 U/muL Cyt
Catalog No. 502908004
Encompass
Exonuclease I (Exo I) fromnbspE. colinbspis an Mg2nbspion dependent exonuclease.
Catalog No. 50-290-8004 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GEE70073X
Exonuclease I (Exo I) fromnbspE. colinbspis an Mg2nbspion dependent exonuclease.