Learn More
LGC GENOMICS LLC (Lucigen) Exonuclease I, 20,000 U @ 20 U/LAn exonuclease that digests single-stranded DNA but not double-stranded DNA, in a 3' to 5' direction.
Catalog No. NC2162084
Encompass
Specific: Processively digest ssDNA in a 3 to 5 direction without harming dsDNA - short 3 overhangs in DNA are not digestedFlexible: Active under a wide variety of buffer conditions and can be added directly into most reaction mixesHeat Inactivated: Destroy the enzyme activity by incubating at 80C for 15 minutesExonuclease I is supplied as enzyme only with no reaction buffer included.
This item is not returnable. View return policy
Catalog No. NC2162084 Supplier LGC GENOMICS LLC Supplier No. X40520K
Specific: Processively digest ssDNA in a 3 to 5 direction without harming dsDNA - short 3 overhangs in DNA are not digestedFlexible: Active under a wide variety of buffer conditions and can be added directly into most reaction mixesHeat Inactivated: Destroy the enzyme activity by incubating at 80C for 15 minutesExonuclease I is supplied as enzyme only with no reaction buffer included.