Learn More
Qiagen Beverly Inc EXONUCLEASE III
Catalog No. 50305936
Encompass
Exonuclease, Exonuclease III; Conc. 100,000 U/ul; 50,000 U; incl. 10X buffer
Catalog No. 50-305-936 Supplier Qiagen Beverly Inc Supplier No. X8020L
Exonuclease, Exonuclease III; Conc. 100,000 U/ul; 50,000 U; incl. 10X buffer