Learn More
LGC GENOMICS LLC (Lucigen) Exonuclease III, 25,000 U @ 200 U/LDigest duplex DNA in a 3 to 5 direction from a nick, blunt end, or 3-recessed end, producing stretches of ssDNA.
Catalog No. NC2162054
Encompass
Produce stretches of ssDNA in dsDNA templates containing nicks, blunt ends or recessed 3' endsProtect protruding 3' ends because this exonuclease will not start digestion at protruding 3' overhangsThis product includes both Exonuclease III enzyme and a 10X reaction buffer.
This item is not returnable. View return policy
Catalog No. NC2162054 Supplier LGC GENOMICS LLC Supplier No. EX4425K
Produce stretches of ssDNA in dsDNA templates containing nicks, blunt ends or recessed 3' endsProtect protruding 3' ends because this exonuclease will not start digestion at protruding 3' overhangsThis product includes both Exonuclease III enzyme and a 10X reaction buffer.