Learn More
New England Biolabs, Inc. Exonuclease VII – 1000 units
SDP
Catalog No. 501047492
Encompass
Exonuclease VII, (Exo VII) derived from E. coli, cleaves single-stranded DNA (ssDNA) from both 5' to 3' and 3' to 5' direction. This enzyme is not active on linear or circular dsDNA. It is useful for removal of single stranded oligonucleotide primers from a completed PCR reaction when different primers are required for subsequent PCR reactions. Digestion of ssDNA by Exonuclease VII is metal-independent.
Catalog No. 50-104-7492 Supplier New England Biolabs, Inc. Supplier No. M0379L
Exonuclease VII, (Exo VII) derived from E. coli, cleaves single-stranded DNA (ssDNA) from both 5' to 3' and 3' to 5' direction. This enzyme is not active on linear or circular dsDNA. It is useful for removal of single stranded oligonucleotide primers from a completed PCR reaction when different primers are required for subsequent PCR reactions. Digestion of ssDNA by Exonuclease VII is metal-independent.