Learn More
System Biosciences LLC ExoQuick Exosome Isolation and RNA Purification Kit (for Serum & Plasma)ExoQuick Exosome Isolation and RNA Purification kit (for Serum & Plasma) = 20 preps
SDP
Catalog No. NC1896951
Encompass_Preferred
Isolate exosomes and then purify exosomal RNAs for biomarker discovery and more (for serum, plasma, or ascites fluid). Looking for biomarkers? Studying exosomal RNAs (exoRNAs)? Turn to SBI’s ExoQuick Exosome Isolation and RNA Purification Kit (for Serum & Plasma) for optimized isolation of exoRNAs. Utilizing ExoQuick for efficient exosome isolation from serum, plasma, or ascites fluid, the kit also contains columns and reagents for purification of RNA from isolated exosomes. For exoRNA isolation from other biofluids, try the ExoQuick Exosome Isolation and RNA Purification Kit (for Media and Urine).
Catalog No. NC1896951 Supplier System Biosciences LLC Supplier No. EQ806A1
Isolate exosomes and then purify exosomal RNAs for biomarker discovery and more (for serum, plasma, or ascites fluid). Looking for biomarkers? Studying exosomal RNAs (exoRNAs)? Turn to SBI’s ExoQuick Exosome Isolation and RNA Purification Kit (for Serum & Plasma) for optimized isolation of exoRNAs. Utilizing ExoQuick for efficient exosome isolation from serum, plasma, or ascites fluid, the kit also contains columns and reagents for purification of RNA from isolated exosomes. For exoRNA isolation from other biofluids, try the ExoQuick Exosome Isolation and RNA Purification Kit (for Media and Urine).