Learn More
System Biosciences LLC EXOQUICK-TC PLUSExoQuick-TC PLUS Exosome Purification Kit - Tissue Culture Media = 10 Reactions
SDP
Catalog No. NC1404480
Encompass_Preferred
Clean and efficient exosome isolation from cell culture media and other biofluids—cleaner and faster than ultracentrifugation, ideal for sensitive applications. While the ExoQuick family of reagents speed-up and simplify exosome isolation and are compatible with most downstream uses, some applications are especially sensitive to carry-over of non-exosomal proteins—applications such as mass spectrometry, Western blotting of low abundance markers, exosome labeling, and in vivo/ex vivo exosome delivery. Which is why SBI offers ExoQuick PLUS.
Catalog No. NC1404480 Supplier System Biosciences LLC Supplier No. EQPL10TC1
Clean and efficient exosome isolation from cell culture media and other biofluids—cleaner and faster than ultracentrifugation, ideal for sensitive applications. While the ExoQuick family of reagents speed-up and simplify exosome isolation and are compatible with most downstream uses, some applications are especially sensitive to carry-over of non-exosomal proteins—applications such as mass spectrometry, Western blotting of low abundance markers, exosome labeling, and in vivo/ex vivo exosome delivery. Which is why SBI offers ExoQuick PLUS.