Learn More
System Biosciences LLC ExoQuick ULTRA EV Isolation SystemExoQuick ULTRA EV Isolation Kit for Serum and Plasma - 20 Reactions
SDP
Catalog No. NC1504033
Encompass_Preferred
The revolutionary new ExoQuick ULTRA is our most advanced polymer-based method for isolating Extracellular Vesicles, resulting in 420X more EVs than ultracentrifugation.Reduces carry-over of albumins by 75% and immunoglobulins by 40% more than other methods, including ultracentrifugation and competitors’ kits. Isolate more EVs per normalized input volume than ultracentrifugation and competitors’ kits. Better biomarker detection—see what you’ve been missing when you increase the sensitivity of EV biomarker detection by up to 11-fold (CD9) and 8-fold (CD81).
Catalog No. NC1504033 Supplier System Biosciences LLC Supplier No. EQULTRA20A1
The revolutionary new ExoQuick ULTRA is our most advanced polymer-based method for isolating Extracellular Vesicles, resulting in 420X more EVs than ultracentrifugation.Reduces carry-over of albumins by 75% and immunoglobulins by 40% more than other methods, including ultracentrifugation and competitors’ kits. Isolate more EVs per normalized input volume than ultracentrifugation and competitors’ kits. Better biomarker detection—see what you’ve been missing when you increase the sensitivity of EV biomarker detection by up to 11-fold (CD9) and 8-fold (CD81).