Learn More
System Biosciences LLC EXOQUICK EXOSOME PRECIPITATION - 5 RXNSExoQuick exosome precipitation solution - 5ml
SDP
Catalog No. NC9023482
Encompass_Preferred
ExoQuick, enables high-throughput, quantitative isolation of exosomes from low volumes (as little as 100 µl) of serum, plasma, or ascites fluid. Need exosomes? SBI's ExoQuick, original formulation, enables high-throughput, quantitative isolation of exosomes from low volumes (as little as 100 µl) of serum, plasma, or ascites fluid. Compatible with a wide variety of downstream applications, ExoQuick is an effective and proven alternative to ultracentrifugation.
Catalog No. NC9023482 Supplier System Biosciences LLC Supplier No. EXOQ5A1
Add to Cart
Add to Cart
ExoQuick, enables high-throughput, quantitative isolation of exosomes from low volumes (as little as 100 µl) of serum, plasma, or ascites fluid. Need exosomes? SBI's ExoQuick, original formulation, enables high-throughput, quantitative isolation of exosomes from low volumes (as little as 100 µl) of serum, plasma, or ascites fluid. Compatible with a wide variety of downstream applications, ExoQuick is an effective and proven alternative to ultracentrifugation.