Learn More
Aviva Systems Biology Corp EXOSC10 Antibody - C-terminal region (ARP40225_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40225_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against EXOSC10. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human EXOSC10. Alternate names: p2, p3, p4, RRP6, PMSCL, Rrp6p, PM-Scl, PMSCL2, PM/Scl-100ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57352