Learn More
Aviva Systems Biology Corp EXOSC10 Antibody C-terminal region (ARP40227_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40227_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EXOSC10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C-terminal region region of human EXOSC10. Alternate names: PM-Scl, PM/Scl-100, PMSCL, PMSCL2, RRP6, Rrp6p, p2, p3, p4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57353