Learn More
Aviva Systems Biology Corp EXOSC2 Antibody - N-terminal region (ARP40762_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40762_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against EXOSC2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human EXOSC2. Alternate names: p7, RRP4, Rrp4p, hRrp4pApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57617