Learn More
Aviva Systems Biology Corp EXOSC3 Antibody - C-terminal region (ARP40824_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40824_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against EXOSC3. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human EXOSC3. Alternate names: p10, PCH1B, RRP40, Rrp40p, CGI-102, hRrp-40, bA3J10.7, RP11-3J10.8ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57631