Learn More
Aviva Systems Biology Corp EXOSC3 Antibody - middle region (ARP40231_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40231_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EXOSC3. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human EXOSC3. Alternate names: p10, RRP40, Rrp40p, CGI-102, hRrp-40, hRrp40p, bA3J10.7, RP11-3J10.8ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57355