Learn More
Aviva Systems Biology Corp EXOSC4 Antibody - N-terminal region (ARP40935_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40935_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against EXOSC4. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human EXOSC4. Alternate names: SKI6, p12A, RRP41, Ski6p, RRP41A, Rrp41p, hRrp41pApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-57669