Learn More
Aviva Systems Biology Corp EXOSC6 Antibody - N-terminal region (ARP41087_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41087_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against EXOSC6. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human EXOSC6. Alternate names: p11, EAP4, MTR3, Mtr3p, hMtr3pApplicationIHC, WB. Species reactivity: Cow, Human, Mouse, Pig, Rat
Catalog No. 50-014-57717