Learn More
Aviva Systems Biology Corp EXOSC6 Antibody - N-terminal region (ARP41086_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41086_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EXOSC6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human EXOSC6. Alternate names: p11, EAP4, MTR3, Mtr3p, hMtr3pApplicationWB. Species reactivity: Cow, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-014-57716