Learn More
Aviva Systems Biology Corp EXOSC7 Antibody - N-terminal region (ARP40796_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40796_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against EXOSC7. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human EXOSC7. Alternate names: p8, EAP1, RRP42, Rrp42p, hRrp42pApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53237