Learn More
Aviva Systems Biology Corp EXOSC9 Antibody - N-terminal region (ARP62142_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62142_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EXOSC9. It was validated on Western Blot. . Alternate names: PM/Scl-75, PMSCL1, RRP45, Rrp45p, p5, p6ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-287-585