Learn More
CAPTIVATE BIO LLC Exosome-Depleted FBS
Catalog No. 5000876336
Encompass_Preferred
Captivate Bio s Exosome-Depleted FBS is designed to enhance the accuracy and efficiency of your research by significantly reducing exosome interference For optimal performance over 95% validated depletion and no prep required
Catalog No. 50-008-76336 Supplier CAPTIVATE BIO LLC Supplier No. FBS162XD250
Captivate Bio s Exosome-Depleted FBS is designed to enhance the accuracy and efficiency of your research by significantly reducing exosome interference For optimal performance over 95% validated depletion and no prep required